Learn More
Cell Size and Diffusion Lab
A quick, easy and safe visual demonstration of cell size and diffusion.
Specifications
Class Size | 40 students |
---|---|
For Use With (Application) | Easy and safe visual demonstration of cell size & diffusion |
Includes | Agar, diffusion media, cups, gloves and rulers, dilute base, comprehensive teacher and student guides |
Time Range | 30 minutes |
Includes:
- Agar
- Diffusion media
- Cups, gloves and rulers
- Dilute base
- Comprehensive teacher and student guides
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S63068
![]() |
Neo/Sci Corporation
201015 |
Why Cells Aren'T Big Refill Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
S63067
![]() |
Neo/Sci Corporation
201013 |
Why Cells Aren'T Big Kit |
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Students construct and analyze simulated cell models of various sizes using a specially formulated agar in the process, they learn how cells obtain nutrients and excrete waste. Data obtained by measuring the cell's volume and calculating its surface area is used to form conclusions about the size limitations imposed by diffusion.
Order Info
Class size: 40 students.
Specifications
40 students | |
Easy and safe visual demonstration of cell size & diffusion | |
Agar, diffusion media, cups, gloves and rulers, dilute base, comprehensive teacher and student guides | |
30 minutes |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.