Learn More
Research Products International CorpSupplier Diversity Partner Cell Spreaders, L Shaped, Sterile, 10 per Package, 500 per Case DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 247660
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use. Supplied in sealed pouches, sterile.
Catalog No. 50-476-530