Learn More
Gibco™ Cell Therapy Systems Hibernate™-A CTS™ Medium Non-distribution product as customer accommodation. Green Product
Cell Therapy Systems Hibernate™-A CTS™ Medium
Manufacturer:  Gibco™ A1370501
Catalog No. A1370501
Add to cart
Description
Description
CTS Hibernate-A Medium is a serum-free, nutrient basal medium for the short-term maintenance of postnatal and adult human tissue in ambient CO2 conditions.
Quick and easy shipment of human tissues
CTS Hibernate-A Medium can be used for shipment of human postnatal and adult tissue such as brain tumors, umbilical cords, and foreskin at ambient temperature using a simple three step process:
1) Collect tissue in a conical tube containing CTS Hibernate-A Medium (brain tumor cells require additional supplements).
2) Ship tissues in CTS Hibernate-A Medium within 24 to 72 hours on cold packs.
3) Transfer received tissues into medium of choice and initiate cultivation.
CTS quality
Gibco CTS products are manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified.
Accompanied by documentation to support your regulatory filing
Specific intended-use statements, full documentation traceability, and convenient access to our Drug Master File (DMF) are available.
CTS Hibernate-A Medium can be combined with CTS B-27 Supplements to yield a complete medium.
For other applications, we provide alternative formulations of CTS Hibernate Medium and CTS B-27 Supplements.
For established protocols using Hibernate Medium, access our Gibco Neurobiology Protocol Handbook.
View publications using Gibco neurobiology products at our Gibco CellCite.
Specifications
Specifications
No
1 x
<1 EU/mL
Liquid
With - Sodium pyruvate
Molecular biology grade, research grade
2 to 8°C
2 to 8°C
Neural Cells
Cell Therapy Systems Hibernate™-A CTS™ Medium, 500mL, Liquid, Animal Origin-Free, Chemically Defined, Serum-Free, High Glucose, Sterile, <1EU/mL Endotoxin Level, For Cell Therapy
Cell therapy
Liquid
Cell Therapy Systems Hibernate™-A CTS™ Medium
1 bottle
Sodium pyruvate
Sterile
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.