Learn More
Cell Biolabs IncSupplier Diversity Partner CELL TRANSFRM ASSAY TRIAL SZ  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner CBA130T
The CytoSelect 96-Well Cell Transformation Assay (Soft Agar Colony Formation) quantifies cells with no manual counting. Cells are incubated for 7-8 days, then quantified using a fluorescent dye. Trial size sufficient for 24 wells.
Catalog No. 50-672-654