Learn More
Cell Biolabs Inc CytoSelect™ Cell Viability and Cytotoxicity Assay Kit, 96 assays
Supplier:  Cell Biolabs Inc CBA240
Encompass_Preferred
The CytoSelect™ Cell Viability and Cytotoxicity Assay Kit provides a colorimetric and fluorometric format for measuring cell viability. Live cells are detected with the MTT reagent or the Calcein AM, and dead cells are detected with the EthD-1 reagent (all provided). Saponin, a cell death initiator, is included as a control. Cell viability/cytotoxicity is determined using the colorimetric and fluorometric detection reagents. The assay principle is basic and can be applied to most eukaryotic cell lines, including adherent and non-adherent cells and certain tissues. The kit does not react with bacteria or yeast. The kit is suitable for use with light and fluorescence microscopes, colorimetric and fluorometric multiwell plate scanners, flow cytometers, and other colorimetric or fluorometric detection systems. The kit contains sufficient reagents for the evaluation of 96 assays in a 96-well plate, or 24 assays in a 24-well plate.
Catalog No. 50-672-679