Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
BioVision IncSupplier Diversity Partner VisionBlue™ Quick Cell Viability Fluorometric Assay Kit: The Easiest & Most Sensitive Fluorometric Assay to Measure Cell Proliferation in response to growth factors, cytokines, mitogens and nutrients etc. Kit Size: 500 & 2500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K303500
The VisionBlue™ Quick Cell Proliferation Assay Kit provides by far the most sensitive and easiest means for quantifying cell proliferation and viability. Simply add the single reagent to cell culture, incubate and read the fluorescence intensity. The VisionBlue™assay utilizes the redox dye resazurin which is not fluorescent, but upon reduction by viable metabolically active cells, the dye becomes highly fluorescent (Ex = 530-570 nm; Em = 590-620 nm). Therefore, viable metabolically active cells can be easily measured. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9679