Learn More
InVitriaSupplier Diversity Partner CELLASTIM REC HSA ALBUMIN 100G DFS Item
Manufacturer:  InVitriaSupplier Diversity Partner 777HSA017100G
Cellastim is recombinant human serum albumin that is completely animal component free. Cellastim has been shown to increase the performance of a wide range of cell types, and is often used as a tool in creating an animal-free cell culture production system. Cellastim helps to facilitate the removal of animal-derived supplements including serum, bovine serum albumin, and human serum albumin without sacrificing performance. In culture that is already animal-free, Cellastim is an important addition to all phases of the culture process from cell cloning and selection to expansion and commercial manufacturing. As a recombinant albumin, Cellastim provides consistent performance compared to animal-derived supplements. Cellastim can be used in chemically defined media or with plant hydrolysates for optimal performance. With the many benefits Cellastim provides, it is a very cost-effective way to maximize cell culture performance.
Catalog No. NC0823944