Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Collagen Type 1-Coated Multiwell Plates
Suitable for serum-free and serum-reduced cell cultivation and transfection
Supplier: Greiner Bio-One 662950
Description
- Coated with Collagen 1
- Improved adhesion and proliferation of primary cells, various cell lines
- Sterile
Specifications
Clear | |
Polystyrene | |
Flat | |
Clear | |
Polystyrene | |
Non-sterile | |
Multi-well Cell Culture Plate | |
24 | |
50 | |
Room Temperature | |
127.76 mm | |
19 mm |
Closure Included | |
Not Autoclavable | |
3.3 mL | |
1.9 sq. cm | |
Rectangular | |
Collagen Type I Protein | |
0.5 to 1.0 mL | |
Lid | |
5 | |
128 x 85 x 19 mm | |
85.48 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction