Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLCOAT™ Collagen Type 1-Coated Multiwell Plates

Suitable for serum-free and serum-reduced cell cultivation and transfection

Manufacturer:  Greiner Bio-One 657950

 View more versions of this product

Catalog No. 07-000-184


Add to cart

Description

Description

  • Coated with Collagen 1
  • Improved adhesion and proliferation of primary cells, various cell lines
  • Sterile
Specifications

Specifications

Clear
Flat
Rectangular
Polystyrene
9.6 sq. cm
Non-sterile
Collagen Type I Protein
16.1 mL
127.76 mm
5
2 to 5 mL
85.48 mm
Not Autoclavable
19 mm
Clear
Closure Included
128 x 85 x 19 mm
Polystyrene
Multi-Well Plate
Lid
50
Room Temperature
6
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit