Learn More
Greiner Bio-One CELLCOAT™ Collagen Type 1-Coated Multiwell Plates
Suitable for serum-free and serum-reduced cell cultivation and transfection
Manufacturer:  Greiner Bio-One 665950005
 View more versions of this product
Catalog No. 07-001-019
Add to cart
Description
Description
  • Coated with Collagen 1
  • Improved adhesion and proliferation of primary cells, various cell lines
  • Sterile
Specifications
Specifications
Clear
Sterile
Multi-Well Plate
5
Cell culture plate
Collagen Type I Protein
Serum-free, serum-reduced cell cultivation and transfection
12