Learn More
Greiner Bio-One CELLCOAT™ Collagen Type 1-Coated Multiwell Plates
Suitable for serum-free and serum-reduced cell cultivation and transfection
$114.10 - $805.50
Specifications
Surface Treatment Collagen Type I Protein
Color Clear
View More Specs
Products 3
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
Catalog Number Mfr. No. Well Count Price Quantity & Availability  
07-000-181
View Documents
Greiner Bio-One
662950
24
Case of 50 for $805.50
Add to cart
07-000-184
View Documents
Greiner Bio-One
657950
6
Case of 50 for $700.00
Add to cart
07-001-019
View Documents
Greiner Bio-One
665950005
12
Case of 5 for $114.10
Add to cart
Description
Description
  • Coated with Collagen 1
  • Improved adhesion and proliferation of primary cells, various cell lines
  • Sterile
Specifications
Specifications
Collagen Type I Protein
Clear