Learn More
Greiner Bio-One CELLCOAT™ Collagen Type I Cell Culture Dishes
For cultivation in serum-free and serum reduced medium. Greiner Bio-One CELLCOAT Collagen Type I Cell Culture Dishes are used for studies of cellular differentiation, cell-matrix interactions, receptor-ligand binding, gene expression and regulation.
$433.50 - $580.00
Specifications
Sterility Non-sterile
Surface Treatment Collagen I
View More Specs
Products 2
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Volume (Metric) Working Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Height (Metric) Volume (Metric) Working Price Quantity & Availability  
07-000-588 View Documents Greiner Bio-One
628950
60 mm 15 mm 6 to 7 mL Case of 100 for $580.00
Add to cart
07-000-685 View Documents Greiner Bio-One
664950
100 mm 20 mm 16 to 17 mL Case of 40 for $433.50
Add to cart
Description
Description
  • Improvement of cell attachment and spreading
  • Cultivation in serum-free and serum reduced medium
  • Culture of primary and transfected cells
  • Shelf life: 24 months at room temperature
Specifications
Specifications
Non-sterile
Collagen I
For research use only. Not for use in diagnostic procedures, unless otherwise indicated