Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Greiner Bio-One CELLCOAT™ Collagen Type I Coated Cell Culture Flask
Ideal for cell cultivation in serum-free or serum-reduced medium
$945.00 - $1980.00
Specifications
Closure Color | Red |
---|---|
Closure Material | HDPE |
Material | Polystyrene |
Neck Style | Canted |
Surface Coating | Collagen Type I Protein |
Catalog Number | Mfr. No. | Culture Area | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Culture Area | Price | Quantity & Availability | |||||
07-000-196
|
Greiner Bio-One
661950 |
175 cm2 |
Case of 40 for $1,980.00
|
|
|||||
07-000-197
|
Greiner Bio-One
690950 |
25cm2 |
Case of 50 for $945.00
|
|
|||||
07-000-653
|
Greiner Bio-One
658950 |
75 cm2 |
Case of 50 for $1,133.00
|
|
|||||
Description
- Promotion of cell adhesion, proliferation and growth of endothelial cell, hepatocytes, muscle cells, pheochromocytoma cells (PC12) and other cell types
- Source: Rat tail Collagen Type I
- Ready-to-use
- Supplied with red filter screw cap.
Specifications
Red | |
Polystyrene | |
Collagen Type I Protein | |
Filter Screw Cap |
HDPE | |
Canted | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title