Learn More
Corning™ CellCube™ Modules
The CellCube™ module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.
$730.72 - $1304.61
Catalog Number | Mfr. No. | Capacity (Metric) | Tray Layers | Surface Treatment | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Tray Layers | Surface Treatment | Price | Quantity & Availability | |||
10-320-201
|
Corning™
3200 |
600 mL | 10 | Tissue Culture Treated |
Case of 2 for $1,304.61
|
|
|||
10-320-202
|
Corning™
3201 |
1.5 L | 25 | Tissue Culture Treated |
Case for $730.72
|
|
|||
10-320-203
|
Corning™
3304 |
1.5 L | 25 | CellBIND |
Case for $1,074.00
|
|
|||
Description
CellCube Modules provide:
- a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
- polystyrene growth surface optimally treated for the growth of attachment dependent cells.
- a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
- an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
Specifications
1 | |
Square | |
Sterile |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.