Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CellEvent™ Caspase-3/7 Green Detection Reagent Available on GSA/VA Contract for Federal Government customers only.

CellEvent™ Caspase-3/7 Green Detection Reagent is a novel fluorogenic substrate for activated caspase-3/7 that is compatible with both live cell and fixed imaging, with absorption/emission maxima of ∼502/530 nm.

$632.00

Specifications

Color Green
Detection Method Fluorescent
Format Vial(s)
Sample Volume 100 µL
Shipping Condition Room Temperature
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
C10423 Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
View Documents
Invitrogen™
C10423
100 µL
Each for $632.00
Add to cart
 
Description

Description

Activation of caspase-3 is an essential event during apoptosis, making this an optimized reagent for analysis of apoptotic cells.

• Optimized caspase-3/7 substrate for apoptosis analysis
• Easy to use—add and read; no cell lysis required
• Use for time course measurements; easily select the time point of interest
• Compatible with both live cell fluorescence-imaging and formaldehyde-based fixation methods
• Multiplex enabled—combine with other fluorescent reagents to confirm apoptosis in the same cell or cell population

Substrate specifics
CellEvent Caspase-3/7 Green Detection Reagent is a four-amino acid peptide (DEVD) conjugated to a nucleic acid-binding dye with absorption/emission maxima of ∼502/530 nm. The DEVD peptide sequence is a cleavage site for caspase-3/7, and the conjugated dye is non-fluorescent until cleaved from the peptide and bound to DNA.

Principles of action
CellEvent Caspase-3/7 Green Detection Reagent is intrinsically non-fluorescent as the DEVD peptide inhibits the ability of the dye to bind to DNA. However, after activation of caspase-3/7 in apoptotic cells, the DEVD peptide is cleaved, enabling the dye to bind to DNA and produce a bright, fluorogenic response. The fluorescent emission of the dye when bound to DNA is ∼530 nm and can be observed using a standard FITC filter set.

Simple three-step protocol
To use CellEvent Caspase 3/7 Detection Reagent, simply add it to cells, incubate 30 minutes, and visualize (see figure). Apoptotic cells with activated caspase-3/7 will have bright green nuclei, while cells without activated caspase 3/7 will have minimal fluorescent signal (see figure).

Assay versatility allows for live cell detection and fixation
This robust assay enables the examination of caspase-3/7 activation in live cells. Furthermore, since wash steps are not necessary for detection, fragile apoptotic cells typically lost during wash steps are preserved. Importantly, the fluorescent signal from CellEvent Caspase 3/7 Detection Reagent can survive fixation and permeabilization. This allows for the flexibility to perform end-point assays and probe for other proteins of interest using immunocytochemistry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Green
Vial(s)
Room Temperature
Fluorescent
100 µL
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit