Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ CellLight™ Nucleus-CFP, BacMam 2.0

CellLight™ Nucleus-CFP, BacMam 2.0

Manufacturer:  Molecular Probes™ C10616

Encompass

Catalog No. C10616


Add to cart

Description

Description

CellLight™ Nucleus-CFP, BacMam 2.0, provides an easy way to label nuclei with cyan fluorescent protein (CFP) in live cells. Simply add the reagent to your cells, incubate overnight, and the cells are ready to image in the morning.

Want to label other cell structures? Learn more about CellLight™ fluorescent protein labeling tools

This ready-to-use construct is transfected into cells using BacMam 2.0 technology, where it expresses CFP fused to the SV40 nuclear localization sequence. You can observe nucleus-CFP behavior in live cells without the cellular toxicity associated with intercalators and label with multiple tracking or tracing dyes to image dynamic cellular processes.

Cells expressing CellLight™ constructs can also be fixed with formaldehyde for multiplexed imaging using immunocytochemical techniques.

CellLight™ Technology is:
  • Fast and convenient: simply add CellLight™ reagent to your cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Less toxic: CellLight™ reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling

    BacMam Technology

    CellLight™ Nucleus-CFP, BacMam 2.0, is a fusion construct of SV40 nuclear localization sequence and CFP, providing accurate and specific targeting to cellular nucleus-CFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight™ reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.
  • Specifications

    Specifications

    Blue
    CFP (Cyan Fluorescent Protein)
    1 vial(s)
    Visible
    CellLight™, Molecular Probes™
    Fluorescence Intensity
    Nucleus
    Store at 2°C to 6°C, protected from light. Do Not Freeze.
    Fluorescence Microscope, Flow Cytometer
    Fluorescent
    Liquid
    Wet Ice
    Fluorescent Protein
    Nucleus
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit

    For Research Use Only. Not for use in diagnostic procedures.