Learn More
Cellpath Acid Resistant Polyester Biopsy Bags
Holds biopsy sample during processing. CellPath Acid Resistant Polyester Biopsy Bags are great alternatives to BioWrap paper and foam biopsy pads. Particularly helpful when collecting endometrial samples from formalin as it can be used as a sieve.
Manufacturer:  Cellpath EBF030002P
 View more versions of this product
Catalog No. 22-222-014
Add to cart
Description
Description
  • Easy tear for quick sample retrieval
  • Acid resistant polyester useful when processing very fine bone samples
  • Useful for collecting endometrial samples from formalin
  • Time saving
Specifications
Specifications
Polyester
100/Pk.
75 x 45mm