Learn More
Cellpath Acid Resistant Polyester Biopsy Bags
Holds biopsy sample during processing. CellPath Acid Resistant Polyester Biopsy Bags are great alternatives to BioWrap paper and foam biopsy pads. Particularly helpful when collecting endometrial samples from formalin as it can be used as a sieve.
$81.20 - $92.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
22-222-013
|
Cellpath
EBF010002P |
40 x 30mm |
Pack of 100 for $81.20
|
|
22-222-014
|
Cellpath
EBF030002P |
75 x 45mm |
Pack of 100 for $92.00
|
|
Description
- Easy tear for quick sample retrieval
- Acid resistant polyester useful when processing very fine bone samples
- Useful for collecting endometrial samples from formalin
- Time saving
Specifications
Polyester | |
100/Pk. |