CellPath Black Diamond™ Disposable Pathology Knives

Well proven, safe alternative to re-sharpening autopsy knives. CellPath Black Diamond™ Disposable Pathology Knives come in a range of five sizes which have high quality Sheffield Stainless Steel double honed blade and molded handle with finger guard.

Manufacturer: CellPath CAA120101A

 View more versions of this product

Catalog No. 22-222-044

  Check Availability
Add to Cart

    Specifications

    Blade Material Sheffield Stainless Steel
    Disposable Yes
    Length (Metric) 30.4cm
    Reusable No
    Product Type Brain Knife
    Length (English) 12 in.

    • Features Sheffield Stainless Steel double-honed blades
    • Molded handle with finger guard for added safety
    • Complete disposal of knife and handle eliminates problems of replacement blade fit and ensures contamination-free environment
    • Available in five different types and lengths (cartilage, trimming, macro, lung and brain knives) to enable pathologist to easily select optimum size of knife for any purpose
    • Supplied in packs of 10 or an assorted pack containing two knives of each type and length

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit