Learn More
Cellpath Black Diamond™ Disposable Pathology Knives
Well proven, safe alternative to re-sharpening autopsy knives. CellPath Black Diamond™ Disposable Pathology Knives come in a range of five sizes which have high quality Sheffield Stainless Steel double honed blade and molded handle with finger guard.
Manufacturer:  Cellpath CAA080101A
 View more versions of this product
Catalog No. 22-222-042
Add to cart
Description
Description
  • Features Sheffield Stainless Steel double-honed blades
  • Molded handle with finger guard for added safety
  • Complete disposal of knife and handle eliminates problems of replacement blade fit and ensures contamination-free environment
  • Available in five different types and lengths (cartilage, trimming, macro, lung and brain knives) to enable pathologist to easily select optimum size of knife for any purpose
  • Supplied in packs of 10 or an assorted pack containing two knives of each type and length
Specifications
Specifications
Sheffield Stainless Steel
8 in.
No
Yes
20.3cm
Macro Knife