Learn More
CellPath CELLMAT Gray Edging for Safety Floor Matting
Matting solution with a specially developed open cell type design to trap wax deposits and avoid potential slipping. CellPath CELLMAT Gray Edging for Safety Floor Matting helps to anchor the mat securely to the floor.
Manufacturer:  CellPath XFC010300H
 View more versions of this product
Catalog No. 22-222-001
  Check Availability
Add to cart
Specifications
Description
  • Bespoke safety floor edging
  • Anti-static
  • Solvent – resistant
  • Non-slip
  • Easily cleaned
Description & Specifications
Specifications
2 x 1m
Hall edging