Learn More
Cellpath CELLMAT Gray Edging for Safety Floor Matting
Matting solution with a specially developed open cell type design to trap wax deposits and avoid potential slipping. CellPath CELLMAT Gray Edging for Safety Floor Matting helps to anchor the mat securely to the floor.
$601.50 - $1166.00
Products 3
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
22-222-000
View Documents
Cellpath
XFC010200B
1.5 x 1 m
Each for $966.00
Add to cart
22-222-001
View Documents
Cellpath
XFC010300H
2 x 1 m
Each for $1,166.00
Add to cart
22-222-002
View Documents
Cellpath
XFC010400D
1 x 1 m
Each for $601.50
Add to cart
Description
Description
  • Bespoke safety floor edging
  • Anti-static
  • Solvent – resistant
  • Non-slip
  • Easily cleaned, wax deposited can be vacuumed out later