Learn More
CellPath CellSafe+ White Biopsy Capsules
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads. CellPath CellSafe+ White Biopsy Capsules features two hinged interlocking frames with an integral nylon mesh that ensures high level of specimen security, zero tissue damage, and low reagent carry-over.
Manufacturer:  CellPath EBE020102A
 View more versions of this product
Catalog No. 22-222-005
  Check Availability
Add to cart
Specifications
Description
  • Features two hinged interlocking frames with an integral nylon mesh
  • Adjustable to biopsy specimen size
  • Hinged in either direction and closed to encapsulate the tissue
  • X–Ray transparent mesh for breast work
  • Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
  • Zero Tissue damage or artefacts associated with foam pads
  • Substantial time savings compared to folding biopsy in tissue paper
Description & Specifications
Specifications
100/Pk.
White