Learn More
Cellpath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes
Provides an innovative solution for the processing and embedding of large tissue specimens. CellPath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes feature an internal chamber a third of the depth of the conventional Supa Mega Cassette.
Manufacturer:  Cellpath EAO011002A
 View more versions of this product
Catalog No. 22-222-127
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Allows use of standard tissue processing cassette to clearly print 2D barcode and test infomation
  • Designed using a docking area for standard processing cassette
  • Strong Hex pore structure improves processing for large specimens
  • Reduces turn around times
  • Time saving by ensuring rapid casting of block, 30 minutes at -8°C
  • Thinner chamber depths reduce specimen thickness and improves specimen quality by reducing distortion with no curling
  • Better sectioning quality through adhesion of paraffin block to cassette
  • Available in various colors
Specifications
Specifications
500/Pk.
Red