Learn More
Cellpath CellPath Stainless-Steel Supa Mega Base Moulds
Reusable base moulds. CellPath Stainless-Steel Supa Mega Base Moulds have extra deep side walls allowing wax to be decanted into the cassette during embedding without the risk of the usual ‘leakage’ of wax.
Manufacturer:  Cellpath GBC361405A
 View more versions of this product
Catalog No. 22-222-031
Add to cart
Description
Description
  • Reusable
  • Stackable to reduce storage area
Specifications
Specifications
5/Pk.
Size: 36 x 36 x 10mm