Learn More
CellPath Supa Mega Tissue Processing Cassettes
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
Manufacturer:  CellPath EAG012002A
 View more versions of this product
Catalog No. 22-222-029
  Check Availability
Add to cart
Specifications
Description
  • Plastic hinged lid
  • Rigid, durable
  • Excellent writing surfaces
  • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
  • Available in a range of colors
Description & Specifications
Specifications
100/Pk.
Turquoise