Learn More
Cellpath Supa Mega Tissue Processing Cassettes
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
Supplier: Cellpath EAG010502A
This item is not returnable.
View return policy
Description
- Plastic hinged lid
- Rigid, durable
- Excellent writing surfaces
- Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
- Available in a range of colors
Specifications
100/Pk. | |
Gray |