CellPath Supa Mega Tissue Processing Cassettes
Widely adopted in research and toxicology laboratories. CellPath Supa Mega Tissue Processing Cassettes are ideal for embedding and processing larger tissue samples of prostrate, brain, breast and eye.
Manufacturer: CellPath EAG011902A
 View more versions of this product
Catalog No. 22-222-028
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Color Lavender
    Quantity 100/Pk.
    • Plastic hinged lid
    • Rigid, durable
    • Excellent writing surfaces
    • Internal chamber is three times depth of the Supa Mega Slim and Supa Mega Mother Ship
    • Available in a range of colors
  • Description & Specifications
Related Products
Product Content Feedback