Learn More
Cellpath System II Hex™ Tissue Processing Cassettes
To hold tissue specimens during embedding, processing, and storage. CellPath System II Hex Tissue Processing Cassettes feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Supplier: Cellpath EAD011003A
This item is not returnable.
View return policy
Description
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Available in 15 stock colors which color code different types of workload or applications
Specifications
1000/Pk. | |
Red | |
Acetyl Resin | |
Tissue Processing Cassette |