Learn More
CellPath System III Hex™ Biopsy Cassettes
To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Biopsy Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Manufacturer:  CellPath EAJ011610A
 View more versions of this product
Catalog No. 22-222-118
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • For use with biopsies
  • Hexagonal perforations arranged in honeycomb structure to utilize available surface area
  • Unique arrangement increases open area 13% thereby increasing reagent flow
  • Includes a fixed lid
  • Features thick side walls for more rigidity
  • Reduces carry over of chemicals
  • Manufactured from high–grade virgin acetyl resin
  • Available in 15 stock colors, which allow laboratories to color code different types of workload or different applications
  • Also manufactured in bespoke colors at the customer’s request
Description & Specifications
Specifications
Yes
500/Pk.
Dark blue
Acetyl resin