CellPath System III Hex™ Biopsy Cassettes

To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Biopsy Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.

Manufacturer: CellPath EAJ010610A

 View more versions of this product

Catalog No. 22-222-110

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Material Acetyl resin
    Color Blue
    Closure Included Yes
    Quantity 500/Pk.

    • For use with biopsies
    • Hexagonal perforations arranged in honeycomb structure to utilize available surface area
    • Unique arrangement increases open area 13% thereby increasing reagent flow
    • Includes a fixed lid
    • Features thick side walls for more rigidity
    • Reduces carry over of chemicals
    • Manufactured from high–grade virgin acetyl resin
    • Available in 15 stock colors, which allow laboratories to color code different types of workload or different applications
    • Also manufactured in bespoke colors at the customer’s request

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit