Learn More
CellPath System III Hex™ Cassettes for Tissue Processing
To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Tissue Processing Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Manufacturer:  CellPath EAI011910A
 View more versions of this product
Catalog No. 22-222-105
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Description
  • Manufactured from high–grade virgin acetyl resin
  • Includes a fixed lid
  • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
  • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
  • Extra thick side walls increases rigidity and stability for high quality sectioning
  • Available in 15 stock colors which color code different types of workload or applications
Description & Specifications
Specifications
Yes
500/Pk.
Lavender
Acetyl resin