CellPath System III Hex™ Cassettes for Tissue Processing
To hold tissue specimens during embedding, processing, and storage. CellPath System III Hex Tissue Processing Cassettes with lid feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
Manufacturer: CellPath EAI011010A
 View more versions of this product
Catalog No. 22-222-099
    N/A
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 
    Specifications
    Material Acetyl resin
    Color Red
    Closure Included Yes
    Quantity 500/Pk.
    • Manufactured from high–grade virgin acetyl resin
    • Includes a fixed lid
    • Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
    • Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
    • Extra thick side walls increases rigidity and stability for high quality sectioning
    • Available in 15 stock colors which color code different types of workload or applications
  • Description & Specifications
Related Products
Product Content Feedback