Learn More
Greiner Bio-One CELLSTAR™ Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap
Ideal for suspension, spheroid cultures and stem cell aggregate formation. Greiner Bio-One CELLSTAR™ Cell-Repellent Surface Cell Culture Flasks with Filter Screw Cap effectively inhibit cell adhesion.
$137.30 - $498.00
Specifications
Material Polystyrene
Surface Coating Cell Repellent
Closure Color White
Closure Type Filter Screw Cap
View More Specs
Products 4
Catalog Number Mfr. No. Volume (Metric) Working Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Working Price Quantity & Availability  
07-000-657
View Documents Promotion Details
Greiner Bio-One
658985
250 mL
Case of 15 for $498.00
Add to cart
07-000-663
View Documents Promotion Details
Greiner Bio-One
660985
550 mL
Case of 5 for $159.20
Add to cart
07-000-668
View Documents Promotion Details
Greiner Bio-One
661985
650 mL
Case of 4 for $137.30
Add to cart
07-000-769
View Documents Promotion Details
Greiner Bio-One
690985
50 mL
Case of 20 for $359.00
Add to cart
Description
Description
  • Made of polystyrene
  • Used for applications such as Gel-based 3D cultures and Magnetic 3D cell culturing (Nano3D technology)
  • Cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments
Specifications
Specifications
Polystyrene
Cell Repellent
White
Filter Screw Cap