Learn More
Greiner Bio-One CELLSTAR™ CELLreactor™ Conical Bottom Polypropylene Filter Top Tube
For shaking suspension cell, aerobic bacteria and microorganisms.
$889.50
Specifications
Closure Type Filter Screw Cap
Closure Color Blue
Sterility Sterile
Graduated Yes
Material Polypropylene
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
07-000-493
View Documents
Greiner Bio-One
227245
50mL Case of 500 for $889.50
Add to cart
07-000-199
View Documents
Greiner Bio-One
188240
15mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Can be used as a small bioreactor for suspension and spheroid cell culture
  • Facilitates high number of parallel experiments
  • Flexible working volume from 1 to 35mL
  • Sterile
  • Allows gas exchange
Specifications
Specifications
Filter Screw Cap
Sterile
Polypropylene
Blue
Yes