Learn More
Partec North America Non-Sterile Celltrics Filters, 250/Box
Supplier:  Partec North America 0400422317
Encompass
The Sysmex Celltrics Non-Sterile Filters, 250/Box are cell strainers used for filtering cells and debris from samples prior to analysis. They have a mesh size of 35μm and are made of nylon. Each box contains 250 non-sterile filters that are individually packaged. The filters have a diameter of 30mm and fit onto 50mL centrifuge tubes. They can be used for a variety of applications including washing, harvesting and filtering cells and tissue. The filters provide high-throughput filtration to obtain a homogenous sample.
Catalog No. NC1942323
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.