Cellucap 1.25 oz. Spunbond Polypropylene Lab Coats

Manufacturer: Cellucap 3302EW/M

 View more versions of this product

Catalog No. 19-304-132

  Check Availability
Add to Cart

    Specifications

    Closure Type Velcro
    Material Spunbond Polypropylene
    Size Medium
    For Use With (Application) Splash protection from nonhazardous liquids and dry particulates
    Disposable Yes
    Weight (English) 1.25 oz.
    Weight (Metric) 0.03kg
    Color White

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit