CelluCap Disposable Open Polypropylene Coverall

Manufacturer: Cellucap 55802L

 View more versions of this product

Catalog No. 19-133-816

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ASIN
    Material Spunbond Polypropylene
    Color White
    For Use With (Application) Lightweight splash protection from non-hazardous liquids and dry particulates
    Seam Type Serged
    Size Large
    Size (English) Chest 25 in.
    Disposable Yes
    Size (Metric) Chest 63.5cm
    Closure Type Zipper
    Size (English) Inseam 29 in.
    Size (Metric) Inseam 73.66cm
    Ankle Style Open
    Cuff Style Open

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit