Learn More
Cellucap Water Repellent SMS Coats
Manufacturer:  Cellucap 3503LG
 View more versions of this product
Catalog No. 19-168-280C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Snap
SMS (Spunbound Melt-Blown Synthetic),Spunbond Polypropylene
For dusty areas and superb for splash protection
White
Large
2-ply Spunbond Polypropylene, inner layer of Meltblown