Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellucap Zipper SMS Coveralls

Catalog Number p-3752458
Click to view available options
Size:
Small
4X-Large
Material:
SMS
Spunbond Polypropylene
This item is not returnable. View return policy
This item is not returnable. View return policy
Size 4X-Large
Material Spunbond Polypropylene
Color White
Ankle Style Open
Cuff Style Open
Closure Type Front Zipper
Description 3-ply protection, Spunbond Polypropylene
For Use With (Application) Light cleanup work
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit