Learn More
Cisbio US IncSupplier Diversity Partner CELLUL'ERK PERK ASSAY KIT 10 DFS Item
Manufacturer:  Cisbio US IncSupplier Diversity Partner 64ERKPEH
Ready-to-use, cell-based kinase assays: The Phospho ERK assay kit is a HTRF sandwich assay for sensitive and rapid detection of activated full length ERK kinase in cell lysate. It allows for monitoring kinase and GPCR signaling on cells.
Catalog No. NC0094468