Learn More
Cellulase, Spectrum™
C1152, 9012-54-8
Manufacturer:  Spectrum Chemical Manufacturing CorporationSupplier Diversity Partner C1152500GM
 View more versions of this product
Catalog No. 18-601-529
Add to cart
Description
Description
Spectrum™ Cellulase , an enzyme produced by bacteria or fungi, breaks down the cellulose molecule into polysaccharides or monosaccharides. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Specifications
Specifications
9012-54-8
Poly Bottle
500 g