Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellulose TEAE Capacity:0.72 meq/g, TCI America™

Click to view available options
Quantity:
10g
Color Undesignated
Quantity 10g
MDL Number MFCD00146355
Synonym Triethylaminoethyl Cellulose
Physical Form Crystal-Powder at 20°C
Chemical Name or Material Cellulose TEAE Capacity:0.72 meq/g