Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CELSR2 Polyclonal Antibody
CELSR2 Polyclonal Antibody
Supplier: Thermo Scientific PA550591
Description
CELSR2 Polyclonal Antibody for IHC (P)

Specifications
CELSR2 | |
Polyclonal | |
Unconjugated | |
CELSR2 | |
ADGRC2; adhesion G protein-coupled receptor C2; cadherin EGF LAG seven-pas G-type receptor 2; cadherin EGF LAG seven-pass G-type receptor 2; Cadherin family member 10; cadherin, EGF LAG seven-pass G-type receptor 2; cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila); CDHF10; Celsr2; EGFL2; EGF-like protein 2; EGF-like-domain, multiple 2; epidermal growth factor-like 2; epidermal growth factor-like protein 2; flamingo; Flamingo homolog; flamingo homolog 3; Flamingo1; flamingo-like protein celsr2; FLJ34118; FLJ42737; FLJ45143; FLJ45845; KIAA0279; MEGF3; mfmi1; mKIAA0279; Multiple EGF-like domains protein 3; multiple epidermal growth factor-like domains 3; multiple epidermal growth factor-like domains protein 3; seven-pass transmembrane cadherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1952, 83465 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9HCU4, Q9QYP2 | |
CELSR2 | |
Synthetic peptide corresponding to a region derived from internal residues of human cadherin, EGF LAG seven-pass G-type receptor 2. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction