Learn More
Cen-Med Urine Midstream Collection and Preservation System
SDP
Manufacturer:  Cen-Med 124500
Catalog No. 21-520-110
Add to cart
This item is not returnable. View return policy
Description
Description
Includes:
  • C&S 10mL vacuum tube with Boric Acid Stabilizer
  • Urine Straw Device for midstream collection with 10CM straw for urinalysis
Individually packed in case of 200
Specifications
Specifications
Midstream urine collection
Test Strip, Transfer Pipette
200/Cs.