Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEND1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$436.00

Specifications

Antigen CEND1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP170495
View Documents
Novus Biologicals
NBP170495
100 μL
Each of 1 for $436.00
Add to cart
 
Description

Description

CEND1 Polyclonal specifically detects CEND1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CEND1
Polyclonal
Rabbit
PBS and 2% Sucrose with 0.09% Sodium Azide
51286
Synthetic peptides corresponding to CEND1(cell cycle exit and neuronal differentiation 1) The peptide sequence was selected from the N terminal of CEND1. Peptide sequence MESRGKSASSPKPDTKVPQVTTEAKVPPAADGKAPLTKPSKKEAPAEKQQ.
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
RUO
BM88 antigen, cell cycle exit and neuronal differentiation 1, cell cycle exit and neuronal differentiation protein 1, FLJ90066
CEND1
IgG
Affinity Purified
15 kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit