Learn More
CENP-Q Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
Supplier: Rockland Immunochemicals 200401B48

Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human CENP-Q protein. A BLAST analysis of the full length sequence was used to suggest partial cross-reactivity with CENP-Q based on the following percentage homologies: macaque (91%), horse (76%), bovine (74%), dog (73%), swine (71%), mouse (65%) and rat (60%). Cross-reactivity with CENP-Q from other sources has not been determined.
CENPQ is a subunit of a CENPH-CENPI-associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.Specifications
CENP-Q | |
Polyclonal | |
Unconjugated | |
CENPQ | |
C6orf139; CENPQ; CENP-Q; Centromere protein Q | |
Rabbit | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunofluorescence, Western Blot | |
1.15 mg/mL | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
Q7L2Z9 | |
CENPQ | |
This protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with full-length human CENP-Q recombinant protein. | |
RUO | |
55166 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.