Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CENPA Recombinant Rabbit Monoclonal Antibody (PSH0-09), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547616

Encompass

Catalog No. PIMA547616


Add to Cart

Description

Description

No Cross with Mouse and Rat. Expression varies across the cell cycle, with high levels in G2 phase (at the mRNA level). Positive control: Hela cell lysate, A375 cell lysate. Subcellular location: Nucleus, Chromosome, centromere, kinetochore.

CENPA Recombinant Monoclonal antibody specifically detects CENPA in Bacteria samples. It is validated for Immunocytochemistry, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CENPA
Recombinant Monoclonal
1 mg/mL
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Rabbit
Protein A
RUO
Bacteria
Liquid
Immunocytochemistry, Western Blot
PSH0-09
Unconjugated
CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607
Proprietary sequence
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit