Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CENPF Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500101APC
Description
CENPF Polyclonal antibody specifically detects CENPF in Human, Mouse, Bovine, Equine samples. It is validated for Flow Cytometry, KnockDownSpecifications
CENPF | |
Polyclonal | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Bovine, Equine | |
Purified |
Flow Cytometry, KnockDown | |
APC | |
AH antigen, cell-cycle-dependent 350K nuclear protein, CENF, CENP-F, CENP-F kinetochore protein, centromere protein F, centromere protein F (350/400kD, mitosin), centromere protein F, 350/400ka (mitosin), centromere protein F, 350/400kDa (mitosin), hcp-1, Kinetochore protein CENPF, mitosin, PRO1779 | |
A bacterial fusion protein from the C-terminus of human CENPF. [UniProt# P49454] | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Chromatin Research | |
1063 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction