Learn More
Aviva Systems Biology Corp CENPI Antibody - N-terminal region (ARP54805_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CENPI. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CENPI. Alternate names: CENP-I, FSHPRH1, LRPR1, Mis6ApplicationIF, WB. Species reactivity: Human
Catalog No. 50-014-61323